Anti-FCRL6 Mouse Monoclonal Antibody [clone: 672505]

Supplier: R & D Systems
MAB6745
102886-662EA 594.37 USD
102886-662
Anti-FCRL6 Mouse Monoclonal Antibody [clone: 672505]
Antibodies
Type: Primary
Antigen Symbol: FCRL6
Antigen Name: Fc receptor-like 6
Antigen Synonyms: FcRH6
Clonality: Monoclonal
Clone: 672505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR