Anti-ERBB2 Rat Monoclonal Antibody [clone: 666521]

Supplier: R & D Systems
MAB6744
102886-660EA 616.06 USD
102886-660
Anti-ERBB2 Rat Monoclonal Antibody [clone: 666521]
Antibodies
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 666521
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR