Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 92320]

Supplier: R & D Systems
MAB674
220021-419EA 763.57 USD
220021-419
Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 92320]
Antibodies
Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: 92320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR