Anti-GPR12 Mouse Monoclonal Antibody [clone: 584940]

Supplier: R & D Systems
MAB6738
102886-658EA 577.02 USD
102886-658
Anti-GPR12 Mouse Monoclonal Antibody [clone: 584940]
Antibodies
Type: Primary
Antigen Symbol: GPR12
Antigen Name: G protein-coupled receptor 12
Antigen Synonyms: GPCR12, GPCR21, PPP1R84
Clonality: Monoclonal
Clone: 584940
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR