Anti-SWAP70 Mouse Monoclonal Antibody [clone: 683317]

Supplier: R & D Systems
MAB6735
102886-656EA 577.02 USD
102886-656
Anti-SWAP70 Mouse Monoclonal Antibody [clone: 683317]
Antibodies
Type: Primary
Antigen: SWAP70
Clonality: Monoclonal
Clone: 683317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR