Anti-PTHLH Mouse Monoclonal Antibody [clone: 677939]

Supplier: R & D Systems
MAB6734
102886-654EA 582.8 USD
102886-654
Anti-PTHLH Mouse Monoclonal Antibody [clone: 677939]
Antibodies
Type: Primary
Antigen: PTHLH
Clonality: Monoclonal
Clone: 677939
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR