Anti-IFNGR1 Mouse Monoclonal Antibody [clone: 92101]

Supplier: R & D Systems
MAB6731
220021-417EA 779.48 USD
220021-417
Anti-IFNGR1 Mouse Monoclonal Antibody [clone: 92101]
Antibodies
Type: Primary
Antigen: IFNGR1
Clonality: Monoclonal
Clone: 92101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR