Anti-PCDH15 Mouse Monoclonal Antibody [clone: 740523]

Supplier: R & D Systems
MAB6729
102886-652EA 577.02 USD
102886-652
Anti-PCDH15 Mouse Monoclonal Antibody [clone: 740523]
Antibodies
Type: Primary
Antigen: PCDH15
Clonality: Monoclonal
Clone: 740523
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR