Anti-GPR75 Mouse Monoclonal Antibody [clone: 689705]

Supplier: R & D Systems
MAB6728
102886-650EA 600.15 USD
102886-650
Anti-GPR75 Mouse Monoclonal Antibody [clone: 689705]
Antibodies
Type: Primary
Antigen Symbol: GPR75
Antigen Name: G protein-coupled receptor 75
Antigen Synonyms: GPRchr2, WI31133
Clonality: Monoclonal
Clone: 689705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR