Anti-DGKB Mouse Monoclonal Antibody [clone: 646523]

Supplier: R & D Systems
MAB6727
102886-648EA 577.02 USD
102886-648
Anti-DGKB Mouse Monoclonal Antibody [clone: 646523]
Antibodies
Type: Primary
Antigen: DGKB
Clonality: Monoclonal
Clone: 646523
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR