Anti-CEBPE Mouse Monoclonal Antibody [clone: 695345]

Supplier: R & D Systems
MAB6726
102886-646EA 577.02 USD
102886-646
Anti-CEBPE Mouse Monoclonal Antibody [clone: 695345]
Antibodies
Type: Primary
Antigen: CEBPE
Clonality: Monoclonal
Clone: 695345
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR