Anti-CANT1 Mouse Monoclonal Antibody [clone: 861206]

Supplier: R & D Systems
MAB6720
102886-644EA 577.02 USD
102886-644
Anti-CANT1 Mouse Monoclonal Antibody [clone: 861206]
Antibodies
Simple Western

Type: Primary
Antigen: CANT1
Clonality: Monoclonal
Clone: 861206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR