Anti-ST6GALNAC5 Mouse Monoclonal Antibody [clone: 719526]

Supplier: R & D Systems
MAB6715
102886-640EA 577.02 USD
102886-640
Anti-ST6GALNAC5 Mouse Monoclonal Antibody [clone: 719526]
Antibodies
Type: Primary
Antigen: ST6GALNAC5
Clonality: Monoclonal
Clone: 719526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR