Anti-DLX5 Mouse Monoclonal Antibody [clone: 712216]

Supplier: R & D Systems
MAB6710
102886-636EA 566.89 USD
102886-636
Anti-DLX5 Mouse Monoclonal Antibody [clone: 712216]
Antibodies
Type: Primary
Antigen: DLX5
Clonality: Monoclonal
Clone: 712216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR