Anti-CD8 Mouse Monoclonal Antibody [clone: 569126]

Supplier: R & D Systems
MAB6709
102886-634EA 577.02 USD
102886-634
Anti-CD8 Mouse Monoclonal Antibody [clone: 569126]
Antibodies
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 569126
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR