Anti-ZEB1 Mouse Monoclonal Antibody [clone: 639914]

Supplier: R & D Systems
MAB6708
102886-632EA 594.37 USD
102886-632
Anti-ZEB1 Mouse Monoclonal Antibody [clone: 639914]
Antibodies
Type: Primary
Antigen: ZEB1
Clonality: Monoclonal
Clone: 639914
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR