Anti-TREML2 Rat Monoclonal Antibody [clone: 656906]

Supplier: R & D Systems
MAB6707
102886-630EA 577.02 USD
102886-630
Anti-TREML2 Rat Monoclonal Antibody [clone: 656906]
Antibodies
Type: Primary
Antigen: TREML2
Clonality: Monoclonal
Clone: 656906
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR