Anti-CAV3 Mouse Monoclonal Antibody [clone: 682708]

Supplier: R & D Systems
MAB6706
102886-628EA 577.02 USD
102886-628
Anti-CAV3 Mouse Monoclonal Antibody [clone: 682708]
Antibodies
Type: Primary
Antigen: CAV3
Clonality: Monoclonal
Clone: 682708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR