Anti-GFRA3 Mouse Monoclonal Antibody [clone: 111004]

Supplier: R & D Systems
MAB6701
220021-415EA 763.57 USD
220021-415
Anti-GFRA3 Mouse Monoclonal Antibody [clone: 111004]
Antibodies
Type: Primary
Antigen Symbol: GFRA3
Antigen Name: GDNF family receptor alpha 3
Antigen Synonyms: GDNFR3
Clonality: Monoclonal
Clone: 111004
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR