Anti-CD160 Mouse Monoclonal Antibody [clone: 688327]

Supplier: R & D Systems
MAB6700
102886-624EA 647.88 USD
102886-624
Anti-CD160 Mouse Monoclonal Antibody [clone: 688327]
Antibodies
Type: Primary
Antigen: CD160
Clonality: Monoclonal
Clone: 688327
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR