Anti-DISC1 Mouse Monoclonal Antibody [clone: 685920]

Supplier: R & D Systems
MAB6699
102886-622EA 577.02 USD
102886-622
Anti-DISC1 Mouse Monoclonal Antibody [clone: 685920]
Antibodies
Type: Primary
Antigen: DISC1
Clonality: Monoclonal
Clone: 685920
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR