Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 882302]

Supplier: R & D Systems
MAB6698
102886-620EA 561.11 USD
102886-620
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: 882302]
Antibodies
Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: 882302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR