Anti-IRAK2 Mouse Monoclonal Antibody [clone: 600848]

Supplier: R & D Systems
MAB6690
102886-618EA 577.02 USD
102886-618
Anti-IRAK2 Mouse Monoclonal Antibody [clone: 600848]
Antibodies
Type: Primary
Antigen: IRAK2
Clonality: Monoclonal
Clone: 600848
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR