Anti-CSPG4 Rat Monoclonal Antibody [clone: 546930]

Supplier: R & D Systems
MAB6689
102886-616EA 655.11 USD
102886-616
Anti-CSPG4 Rat Monoclonal Antibody [clone: 546930]
Antibodies
Type: Primary
Antigen: CSPG4
Clonality: Monoclonal
Clone: 546930
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR