Anti-IL12A Rat Monoclonal Antibody [clone: 45806]

Supplier: R & D Systems
MAB6688
102886-614EA 594.37 USD
102886-614
Anti-IL12A Rat Monoclonal Antibody [clone: 45806]
Antibodies
Type: Primary
Antigen: IL12A
Clonality: Monoclonal
Clone: 45806
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR