Anti-TACR1 Mouse Monoclonal Antibody [clone: 694501]

Supplier: R & D Systems
MAB66871
102886-612EA 594.37 USD
102886-612
Anti-TACR1 Mouse Monoclonal Antibody [clone: 694501]
Antibodies
Type: Primary
Antigen: TACR1
Clonality: Monoclonal
Clone: 694501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR