Anti-TACR1 Mouse Monoclonal Antibody [clone: 694549]

Supplier: R & D Systems
MAB6687
102886-610EA 577.02 USD
102886-610
Anti-TACR1 Mouse Monoclonal Antibody [clone: 694549]
Antibodies
Type: Primary
Antigen: TACR1
Clonality: Monoclonal
Clone: 694549
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR