Anti-MYOG Mouse Monoclonal Antibody [clone: 671037]

Supplier: R & D Systems
MAB66861
102886-608EA 600.15 USD
102886-608
Anti-MYOG Mouse Monoclonal Antibody [clone: 671037]
Antibodies
Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: 671037
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR