Anti-MYOG Mouse Monoclonal Antibody [clone: 671038]

Supplier: R & D Systems
MAB6686
102886-606EA 594.37 USD
102886-606
Anti-MYOG Mouse Monoclonal Antibody [clone: 671038]
Antibodies
Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: 671038
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR