Anti-FCER1A Mouse Monoclonal Antibody [clone: 773704]

Supplier: R & D Systems
MAB6678
102886-604EA 594.37 USD
102886-604
Anti-FCER1A Mouse Monoclonal Antibody [clone: 773704]
Antibodies
Type: Primary
Antigen Symbol: FCER1A
Antigen Name: Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
Antigen Synonyms: FCE1A, FcERI
Clonality: Monoclonal
Clone: 773704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR