Anti-STYK1 Mouse Monoclonal Antibody [clone: 484713]

Supplier: R & D Systems
MAB6668
102886-600EA 577.02 USD
102886-600
Anti-STYK1 Mouse Monoclonal Antibody [clone: 484713]
Antibodies
Type: Primary
Antigen: STYK1
Clonality: Monoclonal
Clone: 484713
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR