Anti-LGALS2 Rat Monoclonal Antibody [clone: 747307]

Supplier: R & D Systems
MAB6667
102886-598EA 577.02 USD
102886-598
Anti-LGALS2 Rat Monoclonal Antibody [clone: 747307]
Antibodies
Type: Primary
Antigen: LGALS2
Clonality: Monoclonal
Clone: 747307
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR