Anti-PRKCZ Mouse Monoclonal Antibody [clone: 453513]

Supplier: R & D Systems
MAB6666
102886-596EA 577.02 USD
102886-596
Anti-PRKCZ Mouse Monoclonal Antibody [clone: 453513]
Antibodies
Type: Primary
Antigen: PRKCZ
Clonality: Monoclonal
Clone: 453513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR