Anti-B4GAT1 Mouse Monoclonal Antibody [clone: 724057]

Supplier: R & D Systems
MAB6664
102886-594EA 561.11 USD
102886-594
Anti-B4GAT1 Mouse Monoclonal Antibody [clone: 724057]
Antibodies
Type: Primary
Antigen: B4GAT1
Clonality: Monoclonal
Clone: 724057
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR