Anti-IL3 Mouse Monoclonal Antibody [clone: 662008]

Supplier: R & D Systems
MAB6658
102886-592EA 561.11 USD
102886-592
Anti-IL3 Mouse Monoclonal Antibody [clone: 662008]
Antibodies
Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: 662008
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR