Anti-DCTN1 Mouse Monoclonal Antibody [clone: 705007]

Supplier: R & D Systems
MAB6657
102886-590EA 594.37 USD
102886-590
Anti-DCTN1 Mouse Monoclonal Antibody [clone: 705007]
Antibodies
Type: Primary
Antigen: DCTN1
Clonality: Monoclonal
Clone: 705007
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR