Anti-PTGER2 Mouse Monoclonal Antibody [clone: 607077]

Supplier: R & D Systems
MAB6656
102886-588EA 594.37 USD
102886-588
Anti-PTGER2 Mouse Monoclonal Antibody [clone: 607077]
Antibodies
Type: Primary
Antigen: PTGER2
Clonality: Monoclonal
Clone: 607077
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR