Anti-LY6K Mouse Monoclonal Antibody [clone: 750747]

Supplier: R & D Systems
MAB6648
102886-586EA 633.42 USD
102886-586
Anti-LY6K Mouse Monoclonal Antibody [clone: 750747]
Antibodies
Type: Primary
Antigen: LY6K
Clonality: Monoclonal
Clone: 750747
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR