Anti-LRP5 Mouse Monoclonal Antibody [clone: 747619]

Supplier: R & D Systems
MAB6646
102886-584EA 561.11 USD
102886-584
Anti-LRP5 Mouse Monoclonal Antibody [clone: 747619]
Antibodies
Type: Primary
Antigen: LRP5
Clonality: Monoclonal
Clone: 747619
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR