Anti-TEX19 Rat Monoclonal Antibody [clone: 678803]

Supplier: R & D Systems
MAB6645
102886-580EA 566.89 USD
102886-580
Anti-TEX19 Rat Monoclonal Antibody [clone: 678803]
Antibodies
Type: Primary
Antigen: TEX19
Clonality: Monoclonal
Clone: 678803
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR