Anti-CNTN3 Rat Monoclonal Antibody [clone: 524518]

Supplier: R & D Systems
MAB6644
102886-578EA 561.11 USD
102886-578
Anti-CNTN3 Rat Monoclonal Antibody [clone: 524518]
Antibodies
Type: Primary
Antigen: CNTN3
Clonality: Monoclonal
Clone: 524518
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR