Anti-STK17B Mouse Monoclonal Antibody [clone: 685228]

Supplier: R & D Systems
MAB6640
102886-576EA 584.25 USD
102886-576
Anti-STK17B Mouse Monoclonal Antibody [clone: 685228]
Antibodies
Type: Primary
Antigen: STK17B
Clonality: Monoclonal
Clone: 685228
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR