Anti-BoNT-D Mouse Monoclonal Antibody [clone: 682810]

Supplier: R & D Systems
MAB6639
102886-574EA 594.37 USD
102886-574
Anti-BoNT-D Mouse Monoclonal Antibody [clone: 682810]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: BoNT-D Heavy Chain
Antigen Synonyms:
Clonality: Monoclonal
Clone: 682810
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: C. botulinum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR