Anti-PIK3R3 Mouse Monoclonal Antibody [clone: 668619]

Supplier: R & D Systems
MAB6638
102886-572EA 594.37 USD
102886-572
Anti-PIK3R3 Mouse Monoclonal Antibody [clone: 668619]
Antibodies
Type: Primary
Antigen: PIK3R3
Clonality: Monoclonal
Clone: 668619
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR