Anti-LBP Rat Monoclonal Antibody [clone: 749405]

Supplier: R & D Systems
MAB6635
102886-570EA 763.57 USD
102886-570
Anti-LBP Rat Monoclonal Antibody [clone: 749405]
Antibodies
Type: Primary
Antigen: LBP
Clonality: Monoclonal
Clone: 749405
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR