Anti-CCL3 Mouse Monoclonal Antibody [clone: 769324]

Supplier: R & D Systems
MAB66252
102886-568EA 763.57 USD
102886-568
Anti-CCL3 Mouse Monoclonal Antibody [clone: 769324]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 769324
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR