Anti-CCL3 Mouse Monoclonal Antibody [clone: 769303]

Supplier: R & D Systems
MAB66251
102886-566EA 594.37 USD
102886-566
Anti-CCL3 Mouse Monoclonal Antibody [clone: 769303]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 769303
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR