Anti-CCL3 Mouse Monoclonal Antibody [clone: 755012]

Supplier: R & D Systems
MAB6625
102886-564EA 561.11 USD
102886-564
Anti-CCL3 Mouse Monoclonal Antibody [clone: 755012]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 755012
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR