Anti-CD79B Mouse Monoclonal Antibody [clone: 683023]

Supplier: R & D Systems
MAB6620
102886-562EA 594.37 USD
102886-562
Anti-CD79B Mouse Monoclonal Antibody [clone: 683023]
Antibodies
Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: 683023
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR