Anti-OSMR Rat Monoclonal Antibody [clone: 118125]

Supplier: R & D Systems
MAB662
220021-408EA 633.42 USD
220021-408
Anti-OSMR Rat Monoclonal Antibody [clone: 118125]
Antibodies
Type: Primary
Antigen: OSMR
Clonality: Monoclonal
Clone: 118125
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR